Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135215.1 | NP_001128687.1 | 8 | 2072 | ||
NM_001135216.1 | NP_001128688.1 | 4 | 1568 | |||
NM_001243241.1 | NP_001230170.1 | 4 | 1493 | |||
NM_001243242.1 | NP_001230171.1 | 4 | 1190 | |||
NM_001243243.1 | NP_001230172.1 | 3 | 1092 | |||
NM_001243244.1 | NP_001230173.1 | 3 | 1081 | |||
NM_030899.4 | NP_112161.3 | 4 | 1557 | |||
NM_145909.2 | NP_665916.1 | 8 | 1694 | |||
XM_005249295.1 | XP_005249352.1 | 4 | 1419 | |||
XM_005249296.4 | XP_005249353.1 | 5 | 5694 | |||
XM_011514807.2 | XP_011513109.1 | 6 | 5744 | |||
XM_011514808.2 | XP_011513110.1 | 4 | 5540 | |||
XM_011514809.1 | XP_011513111.1 | 4 | 1404 | |||
XM_011514811.2 | XP_011513113.1 | 4 | 1919 | |||
XM_011514812.2 | XP_011513114.1 | 4 | 1445 | |||
XM_011514813.2 | XP_011513115.1 | 4 | 1908 | |||
XM_017011196.1 | XP_016866685.1 | 6 | 5832 | |||
GenBank mRNA | AF513019.1 | - | 8 | 1694 | ||
AK027060.1 | - | 1 | 583 | |||
AK057702.1 | - | 8 | 1694 | |||
AK095261.1 | - | 8 | 2072 | |||
AK225382.1 | - | 1 | 586 | |||
AL833416.1 | - | 3 | 852 | |||
AL833617.1 | - | 4 | 1569 | |||
BC008490.1 | - | 4 | 1331 | 4248596 | ||
BC065241.1 | - | 3 | 862 | 6016003 |