Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMBR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022458.3 | NP_071903.2 | 8 | 878 | ||
XM_005249555.2 | XP_005249612.1 | 8 | 853 | |||
XM_005249558.2 | XP_005249615.1 | 8 | 853 | |||
XM_017012507.1 | XP_016867996.1 | 8 | 853 | |||
XM_017012508.1 | XP_016867997.1 | 8 | 811 | |||
XM_017012509.1 | XP_016867998.1 | 8 | 853 | |||
XM_017012510.1 | XP_016867999.1 | 6 | 457 | |||
XM_017012511.1 | XP_016868000.1 | 7 | 709 | |||
XM_017012512.1 | XP_016868001.1 | 6 | 517 | |||
XM_017012513.1 | XP_016868002.1 | 7 | 535 | |||
XM_017012514.1 | XP_016868003.1 | 9 | 947 | |||
XM_017012515.1 | XP_016868004.1 | 8 | 853 | |||
XM_017012516.1 | XP_016868005.1 | 7 | 512 | |||
XM_017012517.1 | XP_016868006.1 | 9 | 931 | |||
XM_017012518.1 | XP_016868007.1 | 10 | 1029 | |||
GenBank mRNA | AF402318.1 | - | 8 | 689 | ||
AK291037.1 | - | 8 | 849 | |||
AK299753.1 | - | 7 | 776 | |||
AL833934.1 | - | 5 | 424 | |||
AL834394.1 | - | 8 | 657 | |||
BC017663.1 | - | 8 | 817 | 3883022 | ||
BC063649.1 | - | 10 | 1025 |