Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMBR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022458.3 | NP_071903.2 | 9, 10 | 967 | ||
XM_005249555.2 | XP_005249612.1 | 9, 10 | 942 | |||
XM_005249558.2 | XP_005249615.1 | 9, 10 | 942 | |||
XM_017012511.1 | XP_016868000.1 | 8, 9 | 798 | |||
XM_017012514.1 | XP_016868003.1 | 10, 11 | 1036 | |||
XM_017012516.1 | XP_016868005.1 | 8, 9 | 601 | |||
XM_017012517.1 | XP_016868006.1 | 10, 11 | 1020 | |||
XM_017012518.1 | XP_016868007.1 | 11, 12 | 1118 | |||
GenBank mRNA | AF402318.1 | - | 9, 10 | 778 | ||
AK291037.1 | - | 9, 10 | 938 | |||
AK299753.1 | - | 8, 9 | 865 | |||
AL833934.1 | - | 6, 7 | 513 | |||
BC017663.1 | - | 9, 10 | 906 | 3883022 | ||
BC063649.1 | - | 11, 12 | 1114 |