Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00452 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278674.1 | NP_001265603.1 | 3 | 451 | ||
GenBank mRNA | AK126042.1 | - | 5 | 562 |