Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284306.1 | NP_001271235.1 | 7 | 2223 | ||
NM_001284307.1 | NP_001271236.1 | 9 | 2238 | |||
NM_022473.2 | NP_071918.1 | 8 | 4666 | |||
XM_005254591.2 | XP_005254648.1 | 11 | 4573 | |||
XM_005254592.2 | XP_005254649.1 | 11 | 4312 | |||
XM_017022483.1 | XP_016877972.1 | 10 | 4348 | |||
XM_017022484.1 | XP_016877973.1 | 8 | 4093 | |||
XM_017022485.1 | XP_016877974.1 | 9 | 1972 | |||
GenBank mRNA | AF205632.1 | - | 8 | 4639 | ||
AK127741.1 | - | 6 | 1205 | |||
AK226132.1 | - | 9 | 2146 | |||
AK302735.1 | - | 10 | 2120 | |||
AL831983.1 | - | 7 | 2169 | |||
AL832014.1 | - | 7 | 2447 | |||
AL832352.1 | - | 7 | 2666 | |||
AL832367.1 | - | 6 | 1117 | |||
AL832455.2 | - | 7 | 2579 | |||
AL833301.1 | - | 7 | 2677 | |||
BX648629.1 | - | 7 | 3771 |