Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFTPD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003019.4 | NP_003010.4 | 8 | 807 | ||
XM_011540087.1 | XP_011538389.1 | 8 | 786 | |||
XM_011540088.2 | XP_011538390.1 | 7 | 758 | |||
GenBank mRNA | BC022318.1 | - | 8 | 808 | 4696173 | |
BM985387.1 | - | 1 | 489 | |||
CR541948.1 | - | 7 | 764 | |||
X65018.1 | - | 8 | 935 |