Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031714.3 | NP_001026884.3 | 11 | 2159 | ||
NM_022489.3 | NP_071934.3 | 11 | 2159 | |||
XM_005268004.4 | XP_005268061.1 | 11 | 2246 | |||
XM_005268005.4 | XP_005268062.1 | 11 | 2217 | |||
XM_017021595.1 | XP_016877084.1 | 11 | 2217 | |||
GenBank mRNA | AK025709.1 | - | 2 | 122 | ||
AK225325.1 | - | 2 | 122 | |||
BC008756.2 | - | 1 | 67 | |||
BG335490.1 | - | 4 | 459 | |||
DQ395338.1 | - | 4 | 420 | |||
DQ395339.1 | - | 4 | 420 | |||
DQ395340.1 | - | 4 | 420 |