Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172810.1 | NP_001166281.1 | 8 | 1077 | ||
NM_023036.4 | NP_075462.3 | 8 | 1080 | |||
XM_011525125.2 | XP_011523427.1 | 8 | 1097 | |||
GenBank mRNA | AF250288.1 | - | 8 | 1037 | ||
AJ295276.1 | - | 7 | 1276 | |||
BC033013.1 | - | 7 | 954 | 4826432 | ||
BC039582.1 | - | 8 | 1067 | 5752153 |