Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039780.3 | NP_001034869.1 | 5, 6 | 1045 | ||
NM_001287252.1 | NP_001274181.1 | 5, 6 | 1093 | |||
NM_001287253.1 | NP_001274182.1 | 5, 6 | 1048 | |||
GenBank mRNA | AK131553.1 | - | 5, 6 | 1046 | ||
BC132837.1 | - | 5, 6 | 1027 | 40146859 | ||
BC136836.1 | - | 5, 6 | 1027 | 9020827 | ||
BC144416.1 | - | 5, 6 | 1030 | 9052937 | ||
BC144417.1 | - | 5, 6 | 1075 | 9052938 |