Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRCOL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195520.1 | NP_001182449.1 | 5 | 514 | ||
XM_011534780.2 | XP_011533082.1 | 5 | 514 | |||
XM_011534784.2 | XP_011533086.1 | 4 | 372 | |||
XM_011534785.2 | XP_011533087.1 | 4 | 377 | |||
XM_017018659.1 | XP_016874148.1 | 4 | 490 | |||
XM_017018660.1 | XP_016874149.1 | 4 | 490 | |||
XM_017018661.1 | XP_016874150.1 | 4 | 732 | |||
XM_017018662.1 | XP_016874151.1 | 4 | 475 | |||
GenBank mRNA | AV653338.1 | - | 2 | 90 | ||
BX282338.1 | - | 5 | 465 | |||
DB344052.1 | - | 2 | 355 | |||
JQ012741.1 | - | 5 | 437 | |||
JQ012742.1 | - | 5 | 569 |