Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122681.1 | NP_001116153.1 | 8 | 1110 | ||
NM_001145855.1 | NP_001139327.1 | 8 | 1253 | |||
NM_001145856.1 | NP_001139328.1 | 8 | 1200 | |||
NM_003023.4 | NP_003014.3 | 8 | 1251 | |||
GenBank mRNA | AB000462.1 | - | 8 | 1251 | ||
AB000463.1 | - | 9 | 1430 | |||
AF000936.1 | - | 7 | 990 | |||
AK124073.1 | - | 3 | 1096 | |||
AK126537.1 | - | 3 | 1467 | |||
AK225497.1 | - | 8 | 1112 | |||
AK299996.1 | - | 8 | 1169 | |||
AK312286.1 | - | 8 | 1048 | |||
BC022996.1 | - | 8 | 1106 | 5103986 | ||
U56386.1 | - | 8 | 1198 |