Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122681.1 | NP_001116153.1 | 4 | 386 | ||
NM_001145855.1 | NP_001139327.1 | 4 | 529 | |||
NM_001145856.1 | NP_001139328.1 | 4 | 476 | |||
NM_003023.4 | NP_003014.3 | 4 | 527 | |||
GenBank mRNA | AB000462.1 | - | 4 | 527 | ||
AB000463.1 | - | 5 | 706 | |||
AF000936.1 | - | 3 | 266 | |||
AK225497.1 | - | 4 | 388 | |||
AK299996.1 | - | 4 | 445 | |||
AK304752.1 | - | 4 | 431 | |||
AK307106.1 | - | 5 | 404 | |||
AK312286.1 | - | 4 | 324 | |||
BC022996.1 | - | 4 | 382 | 5103986 | ||
DC391267.1 | - | 4 | 530 | |||
U56386.1 | - | 4 | 474 |