Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022570.4 | NP_072092.2 | 5 | 750 | ||
NM_197947.2 | NP_922938.1 | 6 | 888 | |||
NM_197948.2 | NP_922939.1 | 5 | 769 | |||
NM_197949.2 | NP_922940.1 | 4 | 631 | |||
NM_197950.2 | NP_922941.1 | 4 | 651 | |||
NR_125336.1 | 7 | 975 | ||||
XM_017019822.1 | XP_016875311.1 | 6 | 846 | |||
XM_017019823.1 | XP_016875312.1 | 5 | 611 | |||
GenBank mRNA | AF313468.1 | - | 7 | 752 | ||
AF313469.1 | - | 5 | 613 | |||
AF400595.1 | - | 6 | 701 | |||
AF400596.1 | - | 5 | 563 | |||
AF400597.1 | - | 5 | 582 | |||
AF400599.1 | - | 4 | 464 | |||
AF400600.1 | - | 3 | 312 | |||
AF400601.1 | - | 6 | 744 | |||
AF400602.1 | - | 8 | 838 | |||
AJ312372.1 | - | 5 | 718 | |||
AJ312373.1 | - | 6 | 856 | |||
AK298679.1 | - | 6 | 804 | |||
AK298724.1 | - | 4 | 567 | |||
AK313247.1 | - | 5 | 681 | |||
AY009090.2 | - | 5 | 634 | |||
AY026769.2 | - | 6 | 701 | |||
AY026770.2 | - | 5 | 563 | |||
AY026771.1 | - | 4 | 444 | |||
AY359002.1 | - | 5 | 722 | |||
CR749536.1 | - | 6 | 1957 |