Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOSPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040097.1 | NP_001035186.1 | 2, 3 | 397 | ||
NM_001040098.1 | NP_001035187.1 | 2, 3 | 313 | |||
NM_001040099.1 | NP_001035188.1 | 1, 2 | 496 | |||
NM_023948.4 | NP_076438.1 | 1, 2 | 496 | |||
XM_005250529.4 | XP_005250586.1 | 2, 3 | 318 | |||
XM_005250531.3 | XP_005250588.1 | 2, 3 | 343 | |||
GenBank mRNA | BC005042.1 | - | 1, 2 | 425 | 3689026 | |
BC011653.2 | - | 1, 2 | 477 | 4299899 | ||
BC028475.2 | - | 1, 2 | 431 | |||
BG164079.1 | - | 495 | ||||
BG707362.1 | - | 1, 2 | 553 | |||
BQ923161.1 | - | 371 |