Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022575.3 | NP_072097.2 | 24 | 2582 | ||
NM_080413.2 | NP_536338.1 | 20 | 2150 | |||
GenBank mRNA | AF308801.1 | - | 24 | 2518 | ||
AK055787.1 | - | 20 | 2120 | |||
AL834401.1 | - | 20 | 2070 | |||
BC012422.1 | - | 12 | 1233 | |||
BC021291.2 | - | 10 | 1101 | |||
BC064406.1 | - | 8 | 838 | |||
BC073959.1 | - | 12 | 1652 | 6182570 | ||
BC126431.1 | - | 9 | 934 | |||
BM468030.1 | - | 5 | 555 |