Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSRNP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320559.1 | NP_001307488.1 | 5 | 1211 | ||
NM_001320560.1 | NP_001307489.1 | 5 | 1012 | |||
NM_033027.3 | NP_149016.2 | 5 | 1080 | |||
XM_017007048.1 | XP_016862537.1 | 5 | 4176 | |||
XM_017007049.1 | XP_016862538.1 | 3 | 464 | |||
GenBank mRNA | AB053121.1 | - | 5 | 1057 | ||
AB063301.1 | - | 5 | 972 | |||
BC038949.1 | - | 5 | 1027 | 5547938 |