Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGF2BP2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK057462.1 | - | 2 | 544 | ||
BC137313.1 | - | 2 | 427 | |||
BC137314.1 | - | 2 | 427 |