Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BNC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001717.3 | NP_001708.3 | 1, 2 | 193 | ||
GenBank mRNA | L03427.1 | - | 189 |