Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPS7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282949.2 | NP_001269878.1 | 2, 3 | 300 | ||
NM_001282950.2 | NP_001269879.1 | 3, 4 | 421 | |||
NM_001282952.2 | NP_001269881.1 | 2, 3 | 345 | |||
NM_001308381.1 | NP_001295310.1 | 3, 4 | 421 | |||
NM_022730.3 | NP_073567.1 | 3, 4 | 421 | |||
XM_006712693.1 | XP_006712756.1 | 3, 4 | 478 | |||
XM_011511638.1 | XP_011509940.1 | 2, 3 | 1861 | |||
XM_011511639.1 | XP_011509941.1 | 2, 3 | 294 | |||
XM_011511640.1 | XP_011509942.1 | 2, 3 | 379 | |||
XM_017004716.1 | XP_016860205.1 | 3, 4 | 478 | |||
XM_017004717.1 | XP_016860206.1 | 2, 3 | 379 | |||
GenBank mRNA | AK022674.1 | - | 3, 4 | 352 | ||
AK024273.1 | - | 2, 3 | 294 | |||
AK098334.1 | - | 1 | 758 | |||
AK124133.1 | - | 4 | 420 | |||
AK126326.1 | - | 2, 3 | 1608 | |||
AK222879.1 | - | 3, 4 | 359 | |||
AK307486.1 | - | 2, 3 | 238 | |||
BC091493.1 | - | 3, 4 | 356 | 30387712 | ||
BX333333.2 | - | 3, 4 | 359 | |||
DA754035.1 | - | 3, 4 | 479 |