Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C16orf58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022744.3 | NP_073581.2 | 8 | 895 | ||
XM_017023570.1 | XP_016879059.1 | 6 | 726 | |||
GenBank mRNA | AK000679.1 | - | 4 | 316 | ||
AK023930.1 | - | 8 | 861 | |||
AK092580.1 | - | 1 | 27 | |||
AK222913.1 | - | 8 | 861 | |||
AK296166.1 | - | 7 | 745 | |||
AK296491.1 | - | 5 | 801 | |||
AK304608.1 | - | 9 | 723 | |||
BC000822.1 | - | 2 | 138 | |||
BC009308.2 | - | 8 | 855 | 4301507 |