Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF574 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022752.5 | NP_073589.4 | 2 | 216 | ||
XM_005259159.2 | XP_005259216.1 | 1 | 79 | |||
XM_011527228.2 | XP_011525530.1 | 2 | 409 | |||
XM_011527229.2 | XP_011525531.1 | 2 | 132 | |||
XM_011527230.2 | XP_011525532.1 | 2 | 84 | |||
XM_017027149.1 | XP_016882638.1 | 2 | 552 | |||
GenBank mRNA | AF447880.1 | - | 2 | 160 | ||
AK027895.1 | - | 2 | 119 | |||
AK131369.1 | - | 2 | 552 | |||
AK131418.1 | - | 2 | 702 | |||
BC001184.1 | - | 2 | 105 | 3502564 | ||
DA404303.1 | - | 2 | 216 |