Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEAF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270875.1 | NP_001257804.1 | 2 | 231 | ||
NM_001270876.1 | NP_001257805.1 | 2 | 231 | |||
NM_022756.5 | NP_073593.2 | 2 | 231 | |||
NR_073090.1 | 2 | 231 | ||||
NR_073091.1 | 2 | 231 | ||||
NR_073092.1 | 2 | 231 | ||||
GenBank mRNA | AK021792.1 | - | 2 | 166 | ||
AK225434.1 | - | 2 | 166 | |||
AK225598.1 | - | 2 | 166 | |||
AK308118.1 | - | 2 | 166 | |||
AY007166.1 | - | 2 | 156 | |||
AY211926.1 | - | 2 | 155 | |||
BC016328.1 | - | 2 | 190 | 4102633 | ||
BC056406.1 | - | 2 | 187 | 6471256 | ||
BM840891.1 | - | 2 | 175 | |||
BX538212.1 | - | 2 | 167 | |||
BX640719.1 | - | 2 | 176 | |||
DQ099384.1 | - | 2 | 158 |