Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENGASE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042573.2 | NP_001036038.1 | 13 | 1744 | ||
XM_006722017.3 | XP_006722080.1 | 10 | 1265 | |||
XM_006722018.3 | XP_006722081.1 | 7 | 1328 | |||
XM_017024961.1 | XP_016880450.1 | 12 | 1586 | |||
GenBank mRNA | AF512564.1 | - | 13 | 1736 | ||
AL110283.1 | - | 9 | 1878 | |||
BC024213.2 | - | 3 | 282 |