Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RMND5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288794.1 | NP_001275723.1 | 8 | 968 | ||
NM_001288795.1 | NP_001275724.1 | 7 | 817 | |||
NM_022762.4 | NP_073599.2 | 7 | 828 | |||
XM_005265969.3 | XP_005266026.1 | 6 | 648 | |||
XM_005265971.4 | XP_005266028.1 | 7 | 1395 | |||
XM_005265972.3 | XP_005266029.1 | 6 | 775 | |||
XM_017009727.1 | XP_016865216.1 | 6 | 1439 | |||
XM_017009728.1 | XP_016865217.1 | 6 | 1106 | |||
GenBank mRNA | AK025971.1 | - | 4 | 373 | ||
AK093481.1 | - | 7 | 769 | |||
AK094065.1 | - | 4 | 975 | |||
AK122692.1 | - | 2 | 1691 | |||
AK125337.1 | - | 8 | 915 | |||
AL713670.1 | - | 6 | 691 | |||
AY359092.1 | - | 4 | 1686 | |||
BC009911.2 | - | 7 | 762 | 2820100 | ||
DQ494789.1 | - | 7 | 762 |