Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146213.1 | NP_001139685.2 | 1, 2 | 95 | ||
NM_022771.4 | NP_073608.4 | 1, 2 | 95 | |||
NR_027449.1 | 1, 2 | 95 | ||||
XM_011538681.2 | XP_011536983.1 | 1, 2 | 59 | |||
XM_017019828.1 | XP_016875317.1 | 1, 2 | 43 | |||
GenBank mRNA | AB449901.1 | - | 1, 2 | 20 | ||
AK022147.1 | - | 1, 2 | 43 | |||
AK225221.1 | - | 1, 2 | 24 | |||
AK225747.1 | - | 1, 2 | 41 | |||
AK292306.1 | - | 1, 2 | 26 | |||
AL833639.1 | - | 1, 2 | 34 | |||
BC028352.2 | - | 1, 2 | 43 | 4827246 |