Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022773.2 | NP_073610.2 | 2 | 350 | ||
NR_033645.1 | 2 | 327 | ||||
NR_033646.1 | 2 | 350 | ||||
XM_006720928.2 | XP_006720991.1 | 2 | 623 | |||
XM_006720929.3 | XP_006720992.1 | 2 | 349 | |||
XM_011522614.2 | XP_011520916.1 | 2 | 348 | |||
XM_011522615.1 | XP_011520917.1 | 2 | 348 | |||
XM_011522618.2 | XP_011520920.1 | 2 | 348 | |||
XM_017023575.1 | XP_016879064.1 | 2 | 348 | |||
XM_017023577.1 | XP_016879066.1 | 2 | 624 | |||
XM_017023578.1 | XP_016879067.1 | 2 | 348 | |||
XM_017023579.1 | XP_016879068.1 | 1 | 153 | |||
XM_017023580.1 | XP_016879069.1 | 2 | 622 | |||
XM_017023581.1 | XP_016879070.1 | 2 | 348 | |||
GenBank mRNA | AB075873.1 | - | 2 | 334 | ||
AK022743.1 | - | 2 | 348 | |||
AK025955.1 | - | 2 | 282 | |||
AK125201.1 | - | 2 | 329 | |||
AK225065.1 | - | 2 | 348 | |||
AK225406.1 | - | 2 | 335 | |||
AK294329.1 | - | 2 | 348 | |||
AK294932.1 | - | 2 | 329 | |||
BC010738.1 | - | 1 | 115 | |||
DA200865.1 | - | 2 | 350 |