Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281517.2 | NP_001268446.1 | 4 | 1004 | ||
NM_001319944.1 | NP_001306873.1 | 5 | 1157 | |||
NM_022778.4 | NP_073615.2 | 5 | 1157 | |||
XM_017002105.1 | XP_016857594.1 | 5 | 1157 | |||
GenBank mRNA | AK024038.1 | - | 5 | 1113 | ||
AK299312.1 | - | 4 | 957 | |||
BC064528.1 | - | 5 | 1108 | 5585742 |