Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322340.1 | NP_001309269.1 | 13 | 1795 | ||
NM_001322341.1 | NP_001309270.1 | 14 | 1899 | |||
NM_001322342.1 | NP_001309271.1 | 13 | 1299 | |||
NM_001322343.1 | NP_001309272.1 | 13 | 1358 | |||
NM_022779.8 | NP_073616.6 | 13 | 1795 | |||
NM_138620.1 | NP_619526.1 | 13 | 1795 | |||
NR_136309.1 | 13 | 1795 | ||||
XM_005272207.4 | XP_005272264.1 | 13 | 1482 | |||
XM_006717236.3 | XP_006717299.1 | 13 | 1797 | |||
XM_011518921.2 | XP_011517223.1 | 13 | 1797 | |||
XM_011518922.2 | XP_011517224.1 | 13 | 1797 | |||
GenBank mRNA | AF335568.1 | - | 13 | 1796 | ||
AF335569.1 | - | 13 | 1796 | |||
AF427339.1 | - | 13 | 1796 | |||
AK023695.1 | - | 9 | 868 | |||
AK027002.1 | - | 13 | 1379 | |||
AK027484.1 | - | 13 | 1796 | |||
AK225121.1 | - | 13 | 1350 | |||
AK225350.1 | - | 13 | 1380 | |||
AK302670.1 | - | 13 | 1532 | |||
BC012726.2 | - | 13 | 1421 |