Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCH7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282805.1 | NP_001269734.1 | 10, 11 | 2292 | ||
NM_001282807.1 | NP_001269736.1 | 7, 8 | 2039 | |||
NM_022826.3 | NP_073737.1 | 8, 9 | 2164 | |||
XM_005246773.2 | XP_005246830.1 | 8, 9 | 2187 | |||
XM_017004719.1 | XP_016860208.1 | 7, 8 | 2022 | |||
XM_017004720.1 | XP_016860209.1 | 7, 8 | 1984 | |||
GenBank mRNA | AK022973.1 | - | 8, 9 | 2122 | ||
AK222880.1 | - | 8, 9 | 2114 | |||
AK292836.1 | - | 10, 11 | 2260 | |||
AK302347.1 | - | 7, 8 | 1988 | |||
AK316382.1 | - | 7, 8 | 1996 | |||
AL833651.1 | - | 4, 5 | 1188 | |||
BC003404.1 | - | 4, 5 | 563 | |||
BC065014.1 | - | 8, 9 | 2088 | 6169288 |