Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258372.1 | NP_001245301.1 | 11 | 1633 | ||
NM_001258373.1 | NP_001245302.1 | 12 | 1736 | |||
NM_022827.3 | NP_073738.2 | 12 | 1681 | |||
GenBank mRNA | AK025000.1 | - | 11 | 1530 | ||
AK025622.1 | - | 7 | 922 | |||
AK092731.1 | - | 12 | 1666 | |||
AK125807.1 | - | 13 | 2503 | |||
AK225189.1 | - | 11 | 1530 | |||
AK291805.1 | - | 12 | 1624 | |||
AK310416.1 | - | 11 | 1525 | |||
BC017468.1 | - | 1 | 125 | |||
BC025255.1 | - | 12 | 1569 | 5019010 | ||
BC065526.1 | - | 12 | 1623 | 6149633 | ||
BC111029.1 | - | 11 | 1538 | 5245538 | ||
CR936769.1 | - | 11 | 1551 | |||
DQ238597.1 | - | 11 | 1599 | |||
GQ472220.1 | - | 11 | 1538 |