Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258372.1 | NP_001245301.1 | 12 | 1737 | ||
NM_001258373.1 | NP_001245302.1 | 13 | 1840 | |||
NM_022827.3 | NP_073738.2 | 13 | 1785 | |||
GenBank mRNA | AK025622.1 | - | 8 | 1026 | ||
AK092731.1 | - | 13 | 1770 | |||
AK125807.1 | - | 14 | 2607 | |||
AK291805.1 | - | 13 | 1728 | |||
BC017468.1 | - | 2 | 229 | |||
BC025255.1 | - | 13 | 1673 | 5019010 | ||
BC065526.1 | - | 13 | 1727 | 6149633 | ||
BC111029.1 | - | 12 | 1642 | 5245538 | ||
CR936769.1 | - | 11 | 1975 | |||
GQ472220.1 | - | 12 | 1642 |