Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022830.2 | NP_073741.2 | 7 | 1459 | ||
GenBank mRNA | AB101208.1 | - | 7 | 1373 | ||
AK025920.1 | - | 3 | 422 | |||
AK026000.1 | - | 7 | 1366 | |||
AK292610.1 | - | 7 | 1458 | |||
BC005013.1 | - | 3 | 421 | |||
BC110910.1 | - | 7 | 1348 | 6141937 | ||
BC128263.1 | - | 7 | 1352 | 40118672 | ||
KU178756.1 | - | 3 | 290 | |||
KU178757.1 | - | 3 | 290 |