Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022830.2 | NP_073741.2 | 2, 3 | 429 | ||
GenBank mRNA | AB101208.1 | - | 2, 3 | 343 | ||
AK026000.1 | - | 2, 3 | 336 | |||
AK292610.1 | - | 2, 3 | 428 | |||
BC110910.1 | - | 2, 3 | 318 | 6141937 |