Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022834.4 | NP_073745.2 | 2 | 596 | ||
GenBank mRNA | AK075366.1 | - | 2 | 419 | ||
AK291557.1 | - | 2 | 403 | |||
BC059409.1 | - | 2 | 403 | 30349211 | ||
BU949921.1 | - | 2 | 446 |