Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAG3L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040585.1 | 5 | 670 | |||
NR_040586.1 | 4 | 594 | ||||
GenBank mRNA | AK307290.1 | - | 5 | 650 | ||
AK308931.1 | - | 5 | 452 | |||
BC026058.2 | - | 4 | 560 | 3936696 | ||
CR457306.1 | - | 3 | 336 |