Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLGAP1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024101.1 | 3 | 818 | |||
GenBank mRNA | AI275162.1 | - | 1 | 202 | ||
AK093095.1 | - | 3 | 818 | |||
BC137437.1 | - | 2 | 701 | |||
BX647607.1 | - | 1 | 2761 |