Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032014.2 | NP_114403.1 | 3 | 252 | ||
GenBank mRNA | AF161453.1 | - | 3 | 179 | ||
AK054982.1 | - | 2 | 293 | |||
BC012167.1 | - | 3 | 204 | 4637888 | ||
BC054865.1 | - | 3 | 219 | 6456699 | ||
CA413186.1 | - | 2 | 458 |
Gene Symbol: URGCP-MRPS24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204871.1 | NP_001191800.1 | 6 | 394 |