Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182640.2 | NP_872578.1 | 1 | 107 | ||
GenBank mRNA | AK001451.1 | - | 1 | 49 | ||
AK130078.1 | - | 1 | 48 | |||
AV729381.1 | - | 1 | 84 | |||
BC036575.1 | - | 1 | 60 | |||
BC057240.1 | - | 1 | 61 | 6495350 | ||
DT217261.1 | - | 1 | 107 |