Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321995.1 | NP_001308924.1 | 9, 10 | 923 | ||
NM_001321996.1 | NP_001308925.1 | 9, 10 | 1023 | |||
NM_001321997.1 | NP_001308926.1 | 9, 10 | 844 | |||
NM_031902.4 | NP_114108.1 | 9, 10 | 1102 | |||
NR_136152.1 | 8, 9 | 932 | ||||
NR_136153.1 | 8, 9 | 1011 | ||||
GenBank mRNA | AB049940.1 | - | 9, 10 | 983 | ||
AK058160.1 | - | 8, 9 | 785 | |||
BC000219.2 | - | 5, 6 | 417 | |||
BC014172.2 | - | 9, 10 | 1075 | 4561399 |