Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NBEAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114132.1 | NP_001107604.1 | 50 | 7689 | ||
XM_005246787.3 | XP_005246844.1 | 50 | 7715 | |||
XM_005246788.2 | XP_005246845.1 | 29 | 4191 | |||
XM_006712698.3 | XP_006712761.1 | 51 | 7802 | |||
XM_006712699.3 | XP_006712762.1 | 51 | 7802 | |||
XM_011511658.2 | XP_011509960.1 | 51 | 7757 | |||
XM_011511659.2 | XP_011509961.1 | 51 | 7814 | |||
XM_011511660.2 | XP_011509962.1 | 47 | 7136 | |||
XM_011511662.2 | XP_011509964.1 | 43 | 6471 | |||
XM_017004728.1 | XP_016860217.1 | 38 | 5869 | |||
XM_017004729.1 | XP_016860218.1 | 26 | 3883 | |||
GenBank mRNA | AK127772.1 | - | 22 | 3350 | ||
AK128636.1 | - | 20 | 2416 | |||
AY172970.1 | - | 20 | 2595 | |||
BX647133.1 | - | 14 | 1516 |