Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLA-DRB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001298.1 | 2 | 234 | |||
GenBank mRNA | AF005487.1 | - | 2 | 234 | ||
L76565.1 | - | 2 | 234 | |||
U33547.1 | - | 2 | 254 |