Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278063.1 | NP_001264992.1 | 6 | 2952 | ||
XM_006722525.3 | XP_006722588.1 | 6 | 3293 |