Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHLDB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198850.3 | NP_942147.3 | 8, 9 | 1290 | ||
XM_005259169.3 | XP_005259226.1 | 8, 9 | 1358 | |||
XM_005259170.3 | XP_005259227.1 | 8, 9 | 1358 | |||
XM_005259171.3 | XP_005259228.1 | 8, 9 | 1358 | |||
XM_011527236.2 | XP_011525538.1 | 8, 9 | 1462 | |||
GenBank mRNA | BC157852.1 | - | 2, 3 | 274 | ||
BC171853.1 | - | 2, 3 | 283 | |||
BG403419.1 | - | 2, 3 | 141 |