Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FER1L6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039112.2 | NP_001034201.2 | 29, 30 | 4031 | ||
XM_006716618.3 | XP_006716681.1 | 29, 30 | 3958 | |||
XM_011517231.2 | XP_011515533.1 | 30, 31 | 3997 | |||
XM_011517232.2 | XP_011515534.1 | 30, 31 | 4138 | |||
XM_011517233.2 | XP_011515535.1 | 29, 30 | 4099 | |||
XM_011517235.2 | XP_011515537.1 | 30, 31 | 3998 | |||
XM_011517236.2 | XP_011515538.1 | 10, 11 | 1726 | |||
GenBank mRNA | AB196633.1 | - | 29, 30 | 4031 | ||
DV080599.1 | - | 1, 2 | 115 |