Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323971.1 | NP_001310900.1 | 12 | 1886 | ||
NM_001323972.1 | NP_001310901.1 | 11 | 1583 | |||
NM_001323973.1 | NP_001310902.1 | 12 | 1886 | |||
NM_001323974.1 | NP_001310903.1 | 12 | 1800 | |||
NM_001323975.1 | NP_001310904.1 | 12 | 1800 | |||
NM_004594.2 | NP_004585.1 | 12 | 1806 | |||
NR_136664.1 | 12 | 1800 | ||||
NR_136665.1 | 11 | 1676 | ||||
NR_136666.1 | 10 | 1637 | ||||
NR_136667.1 | 12 | 1800 | ||||
GenBank mRNA | AB209203.1 | - | 11 | 2611 | ||
AF111173.1 | - | 12 | 1812 | |||
AL137689.1 | - | 3 | 157 | |||
BC142671.1 | - | 12 | 1769 | 40148180 | ||
BC144131.1 | - | 11 | 1676 | |||
BC150207.1 | - | 11 | 2062 |