Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166496.1 | NP_001159968.1 | 4 | 1534 | ||
NM_003051.3 | NP_003042.3 | 4 | 1065 | |||
XM_011542026.2 | XP_011540328.1 | 4 | 798 | |||
XM_011542027.2 | XP_011540329.1 | 4 | 808 | |||
GenBank mRNA | AK094064.1 | - | 4 | 1534 | ||
AK296688.1 | - | 4 | 848 | |||
AK303603.1 | - | 4 | 907 | |||
AK312500.1 | - | 4 | 908 | |||
AL162079.1 | - | 4 | 895 | |||
BC026317.1 | - | 4 | 908 | 4811581 | ||
BC045664.1 | - | 5 | 1619 | 5298234 | ||
L31801.1 | - | 3 | 714 |