Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC19A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205207.1 | NP_001192136.1 | 4, 5 | 1203 | ||
NM_194255.2 | NP_919231.1 | 5, 6 | 1454 | |||
XM_005261164.3 | XP_005261221.1 | 5, 6 | 1427 | |||
XM_011529696.2 | XP_011527998.1 | 6, 7 | 1886 | |||
XM_011529698.2 | XP_011528000.1 | 5, 6 | 1490 | |||
XM_011529700.2 | XP_011528002.1 | 5, 6 | 1749 | |||
XM_011529701.2 | XP_011528003.1 | 6, 7 | 1699 | |||
XM_011529702.2 | XP_011528004.1 | 5, 6 | 1413 | |||
XM_011529703.2 | XP_011528005.1 | 5, 6 | 1481 | |||
XM_011529704.2 | XP_011528006.1 | 5, 6 | 1397 | |||
XM_011529706.2 | XP_011528008.1 | 4, 5 | 1842 | |||
XM_011529709.2 | XP_011528011.1 | 5, 6 | 1668 | |||
XM_017028443.1 | XP_016883932.1 | 5, 6 | 1578 | |||
GenBank mRNA | AF004354.1 | - | 4, 5 | 1372 | ||
AK303168.1 | - | 4, 5 | 1190 | |||
AK307623.1 | - | 5, 6 | 1458 | |||
AK313125.1 | - | 5, 6 | 1418 | |||
S78996.1 | - | 4, 5 | 1301 | |||
U15939.1 | - | 5, 6 | 1395 | |||
U17566.1 | - | 5, 6 | 1395 | |||
U19720.1 | - | 5, 6 | 1396 |