Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003057.2 | NP_003048.1 | 1 | 167 | ||
NM_153187.1 | NP_694857.1 | 1 | 167 | |||
XM_005267102.4 | XP_005267159.1 | 1 | 209 | |||
XM_005267103.1 | XP_005267160.1 | 1 | 209 | |||
XM_006715552.1 | XP_006715615.1 | 1 | 209 | |||
GenBank mRNA | AK289887.1 | - | 1 | 125 | ||
AV684761.1 | - | 1 | 164 | |||
BC126364.1 | - | 1 | 133 | 8992080 | ||
U77086.1 | - | 1 | 115 | |||
X98332.1 | - | 1 | 134 |