Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289396.1 | NP_001276325.1 | 14 | 2369 | ||
NM_001289397.1 | NP_001276326.1 | 14 | 2382 | |||
NM_003070.4 | NP_003061.3 | 14 | 2382 | |||
NM_139045.3 | NP_620614.2 | 14 | 2382 | |||
GenBank mRNA | AK296373.1 | - | 14 | 1770 | ||
AK299683.1 | - | 5 | 575 | |||
X72889.1 | - | 14 | 2370 |