Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other STK33 SIRNA Assays ›
Gene Symbol:
STK33
Gene Name:
serine/threonine kinase 33
Gene Aliases:
-
Chromosome Location:
Chr.11: 8335029 - 8594289 on Build GRCh38
UniGene ID:
Hs.501833
Species:
Human
siRNA ID
HSS185954
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: STK33
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001289058.1
NP_001275987.1
7
842
NM_001289059.1
NP_001275988.1
4
549
NM_001289061.1
NP_001275990.1
6
1107
NM_030906.3
NP_112168.1
7
1256
XM_011520288.2
XP_011518590.1
7
2334
XM_011520290.2
XP_011518592.1
9
1253
XM_011520292.2
XP_011518594.1
7
1055
XM_011520293.2
XP_011518595.1
7
1084
XM_011520294.1
XP_011518596.1
6
1049
XM_011520295.2
XP_011518597.1
6
3145
XM_011520297.2
XP_011518599.1
6
822
XM_017018143.1
XP_016873632.1
9
1313
XM_017018144.1
XP_016873633.1
9
1215
XM_017018145.1
XP_016873634.1
8
3247
XM_017018146.1
XP_016873635.1
9
1377
XM_017018147.1
XP_016873636.1
8
3174
XM_017018148.1
XP_016873637.1
7
3106
XM_017018149.1
XP_016873638.1
8
1258
XM_017018150.1
XP_016873639.1
8
1113
XM_017018151.1
XP_016873640.1
8
1072
XM_017018152.1
XP_016873641.1
6
1403
XM_017018153.1
XP_016873642.1
6
1001
XM_017018154.1
XP_016873643.1
6
860
XM_017018155.1
XP_016873644.1
6
1415
XM_017018156.1
XP_016873645.1
6
861
XM_017018157.1
XP_016873646.1
6
626
XM_017018158.1
XP_016873647.1
7
912
XM_017018159.1
XP_016873648.1
8
3250
XM_017018160.1
XP_016873649.1
6
1049
XM_017018161.1
XP_016873650.1
8
3256
XM_017018162.1
XP_016873651.1
7
859
XM_017018163.1
XP_016873652.1
6
850
XM_017018164.1
XP_016873653.1
6
736
XM_017018165.1
XP_016873654.1
7
815
GenBank mRNA
AJ303380.2
-
5
1600
AK092720.1
-
4
488
AK093251.1
-
6
965
AK122808.1
-
3
3175
AK293193.1
-
7
708
BC031231.1
-
7
1204
5296502
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein autophosphorylation
protein serine/threonine kinase activity
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-4vtwv:80/10.239.167.181:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline